Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Bromo-10'-(2-naphthyl)-9,9'-bianthracene 98.0+%, TCI America™
SDP

Catalog No. B48961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48961G 1 g
B4896200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48961G Supplier TCI America Supplier No. B48961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1172087-81-8
Molecular Formula C38H23Br
Molecular Weight (g/mol) 559.506
MDL Number MFCD22987563
InChI Key OUMDUWOVDLPNDY-UHFFFAOYSA-N
PubChem CID 91972118
IUPAC Name 9-bromo-10-(10-naphthalen-2-ylanthracen-9-yl)anthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=C7C=CC=CC7=C(C8=CC=CC=C86)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 559.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 10-Bromo-10′-(2-naphthyl)-9,9′-bianthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.