Learn More
AdipoGen [10]-Gingerol (10 mg)
SDP
Supplier:  AdipoGen CDXG0198M010
Encompass_Preferred
Chemical. CAS: 23513-15-7. Formula: C21H34O4. MW: 350.49. Isolated from plant source. 10-Gingerol is originally found in species of Zingiber. It exhibits antiemetic, anticancer, antioxidative, anti-inflammatory and antibiotic activities. 10-Gingerol increases radical scavenging of superoxide and hydroxyl radicals, inhibits oxidative burst activity and decreases expression of NO and PGE2 in vitro. It exhibits antioxidant behaviour towards lipids and is effective in suppressing obesity and adipose tissue inflammation. 10-Gingerol also inhibits 5-HT3 receptors. Additionally, 10-gingerol inhibits proliferation in different cancer cells. Shwon to suppresses IL-2-induced proliferation of T lymphocytes. This compound also displays antibacterial efficacy against gram negative bacteria such as Porphyromonas and Prevotella, antifungal and antiparasitic activity.
Catalog No. 50-205-4465
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.