Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 10-Hydroxycamptothecin

Catalog No. AC467490010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467490010 1 g
AC467490050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467490010 Supplier Thermo Scientific Chemicals Supplier No. 467490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 19685-09-7 | C20H16N2O5, C20H16N2O5 | 364.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19685-09-7
Molecular Formula C20H16N2O5

Specifications

Quantity 1 g
Melting Point 225.0°C to 267.0°C
Formula Weight 364.36
InChI Key HAWSQZCWOQZXHI-FQEVSTJZSA-N
SMILES CC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC(O)=CC1=C3)C2=O
IUPAC Name (19S)-19-ethyl-7,19-dihydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular Weight (g/mol) 364.36
Chemical Name or Material 10-Hydroxycamptothecin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.