Learn More
Apexbio Technology LLC Anthraquinone-2-carboxylic acid 117-78-2 250mg
Supplier: Apexbio Technology LLC C7037250
Anthraquinone-2-carboxylic acid is a natural anthraquinone derivative structurally characterized by an anthraquinone backbone with a carboxylic acid group at position 2 It modulates specific signaling pathways involved in the inflammatory response and cellular communication thereby regulating inflammation Anthraquinone-2-carboxylic acid exerts its biological activity primarily through modulation of inflammation-related signaling pathways Its bioactivities include anti-inflammatory and antinociceptive effects Based on these pharmacological properties Anthraquinone-2-carboxylic acid holds research potential in the investigation of inflammation mechanisms and pain management
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.