Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ 10 in. Original Roll-O-Matic™ Refills. Galv. BULK

Catalog No. 19200800 Shop All Vileda Professional Products
Change view
Click to view available options
60/case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-200-800 60/case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-800 Supplier Vileda Professional™ Supplier No. 120949

Manufactured in the USA and are ideal for use with the 'Pull and Lift' technique most commonly used in controlled environments.

  • Original refills have been the standard in the professional and controlled environment industry for well over 30 years
  • For use with all Roll-O-Matic™ applications
  • Fully autoclavable
  • Bulk packaged

Specifications

Product Type Original Roll-O-Matic™ Refills
Color Blue
Autoclavable Autoclavable
Size 10 in.
For Use With (Application) All Roll-O-Matic&trade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.