Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 10 μL Microvolume Filter Tips
GSA_VA

Catalog No. 14222784 Shop All Axygen Products
Change view
Click to view available options
For Use With (Equipment):
Universal Fit Micro
Tip Style:
Maxymum Recovery, Universal
Microvolume
Universal, Maxymum Recovery
Universal, Microvolume
Format:
Bulk
Rack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Tip Style Format
14-222-784 Universal Fit Micro Universal, Maxymum Recovery Rack
14-222-783 Universal Fit Micro Universal, Microvolume Bulk
14-222-785 Universal Fit Micro Universal, Microvolume Rack
14-222-792 Universal Fit Micro Maxymum Recovery, Universal Rack
14-222-793 Universal Fit Micro Microvolume Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-222-784 Supplier Axygen™ Supplier No. TF300LRS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Order Info

    • Non-Filtered bulk: 1000 tips/pack, 240 packs/case, Filtered bulk: 1000 tips/pack, 10 packs/case
    • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
    • Stack Racks: 96 tips/rack, 5 racks/stack,10 stacks/case

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 0.5 to 10 μL
    Volume (Metric) Dispensing 0.5 to 10 μL
    Filtered Filtered
    Sterility Sterile
    Color Clear
    Autoclavable Not Autoclavable
    Certifications/Compliance USP 3127
    Clean Claims DNase, Rnase Free
    For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
    Format Rack
    Tip Style Universal, Maxymum Recovery
    Packaging Rack
    Quantity 960/Pk.
    Product Type Filtered Pipette Tips
    Includes 10 Racks x 96 Tips
    Green Features Bulk Handheld Tips Use on Average89% Less Resin Than Their Pre-racked Alternatives.
    Disposable Single-use
    For Use With (Equipment) Universal Fit Micro
    Graduated No
    Material Polypropylene
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.