Learn More
Cayman Chemical (±)-Talinolol, 57460-41-0, 10 mg
SDP
Supplier:  Cayman Chemical 1611610
Encompass_Preferred
Molecular Formula C20H33N3O3, Purity ≥95%, Formula Weight 363.5, Synonym: N-cyclohexyl-N'-[4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-urea; A 1-selective adrenoceptor antagonist well known for its cardioprotective and antihypertensive activity; often used to examine the activity of the drug-transporting MDR1 gene product P-glycoprotein.
Catalog No. NC1048285
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.