Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ 10 mm/10 mm Union PTFE

Catalog No. 111009666 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
3/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-100-9666 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9666 Supplier MilliporeSigma™ Supelco™ Supplier No. 28412U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For connecting glass dioxin columns and reversible tubes with 10 mm stem O.D.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Quantity 3/Pk.
Description For connecting glass dioxin columns and reversible tubes with 10 mm stem O.D.
Material PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.