Learn More
Cayman Chemical 10-Nitrooleate, 875685-46-4, 50 µg
SDP
Supplier:  Cayman Chemical 1000804350
Encompass_Preferred
Nitration product of oleic acid in vivo mediated by peroxynitrite, acidified nitrite, or myeloperoxidase (in the presence of H2O2 and nitrite)
Catalog No. NC1523649
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.