Learn More
DIAGNOX LLC 10 PARAMETER UA TESTS 20/PK
Supplier:  DIAGNOX LLC URINOX1020
Encompass_Preferred
URINOX10-20 Ten Parameter Urinalysis UA Reagent Strips (LEU,NIT, URO, BIL, PRO, PH, KET, GLU, BLO, SG). CLIA Waived, FDA 510k Professional/Prescription Use, Health Canada, CE, UKCA
Catalog No. NC2866619