Learn More
Marathon LS 100-1000 µL Endurance single channel Pipettor
SDP
Supplier:  Marathon LS MLSMP11000
Encompass_Preferred
100-1000μL MarathonLS Endurance Single Channel Pipettor. Extremely low force pipetting plunger. Fully autoclavable and UV resistant. Adjustable tip ejector. Designed for universal fit with all commonbrands of pipet tips. Finger hook and ergonomic anti-fatigue design. Easy in-house calibration. Volume can be set by turning either the thumb wheel or the plunger button. Volume adjustment is protected by a locking mechanism. Factory calibrated to strict ISO 8655. Compatible with MLS-PTFS series.
Catalog No. NC2065666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.