Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 100 cal Arc Flash Kit with Voltage Gloves (09)
SDP

Catalog No. 170011603 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1603 2X-Large
17-001-1583 Small
17-001-1588 Medium
17-001-1593 Large
17-001-1598 X-Large
17-001-1608 3X-Large
17-001-1613 4X-Large
17-001-1618 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1603 Supplier National Safety Apparel™ Supplier No. KIT4SC1002X09
Add to Cart
Add to Cart
Request a Quote

Conveniently packed in one bag, all of the 100 cal protection you need is close at hand. Features a generous fit for easy movement and comfort while staying inherently flame resistant.

  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Hook and loop at side for easy access

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Gender Unisex
Product Type Arc Flash Kit
Material DuPont™ Nomex™, Kevlar™
Arc Flash Rating 100 cal/cm²
Size 2X-Large
Color Brown
Suspender Adjustable
Ankle Style Open
Closure Type FR Hook and Loop
Seam Type Stitched
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.