Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ 100 g-5 mg Analytical Precision UltraClass Weight Set

Catalog No. 02218053 Shop All Troemner Products
Change view
Click to view available options
Certifications/Compliance:
NVLAP Accredited Certificate
No Certificate
Traceable Certificate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
02-218-053 Traceable Certificate
02-218-055 No Certificate
02-218-054 NVLAP Accredited Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-218-053 Supplier Troemner™ Supplier No. 30390111
Add to Cart
Add to Cart

Combine high precision with the advantage of two-piece construction. UltraClass weights, exclusive to Troemner™ are the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

  • Used in extremely stable environments
  • 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weights 1 g and above are of two-piece construction
  • Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Traceable Certificate
Classification UltraClass
Material Troemner Alloy 8, Stainless Steel
Case Material Polycarbonate
Density Weights 1 g and above are 8.03 g/cm3 at 20°C. Weights below 1 g are 7.95 g/cm3 at 20°C.
Mass 100 g to 5 mg
Length (Metric) 20.32 cm
Height (Metric) Case 8.89 cm
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Includes Cylindrical Weight: 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, Sheet Weight: 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, Polycarbonate Case, Forceps, Brush
Quantity 18/Set
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.