Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 100% Graphite Ferrules

Catalog No. 29013487
Change view
Click to view available options
For Use With (Equipment):
For Thermo M4 Nut
Description:
0.25mm I.D.
0.32mm I.D.
0.53mm I.D.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Description
29013487 For Thermo M4 Nut 0.32mm I.D.
29013488 For Thermo M4 Nut 0.25mm I.D.
29013486 For Thermo M4 Nut 0.53mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 29013487 Supplier Thermo Scientific™ Supplier No. 29013487

Composed of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections

  • Easy-to-use
  • Form a soft grip with the column and provide a stable seal

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) For Thermo M4 Nut
Description 0.32mm I.D.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.