Learn More
Market Lab 50mL One Hundred Place Tube Rack
Supplier:  Market Lab 35859
Encompass_Preferred
The 50mL One Hundred Place Tube Rack by MarketLab is a plastic rack that holds up to 100 tubes with diameters of 12 to 17mm. It has numbered positions from 1 to 100 for easy organization and identification. The rack measures 8 inches wide by 12 inches deep by 2 inches high. It is made from crystal polystyrene for chemical resistance and can withstand temperatures from -20 to 121 degrees Celsius. This tube rack provides an organized storage solution for various lab tubes.
Catalog No. NC2171565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.