Description
- Provides excellent sorbency
- Sterilizable by autoclave, ethylene oxide, or gamma radiation
- Can be used with fiberglass handle
Specifications
Specifications
| Product Type | Mop Head |
| Head Type | String |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Color-coded | Yes |
| Material | Non-woven Cellulose, Polyester |