Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

100 ppb TO-15 65 Component Mix Gas Standard in Nitrogen, Restek
SDP

Catalog No. 06611920 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
110 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-611-920 110 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-920 Supplier Restek Corporation Supplier No. 26360
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

  • High-quality air monitoring gas calibration standards provided by Spectra/Linde and Airgas (formerly Scott/Air Liquide gas)
    • Meets lab requirements for two separate sources of calibration standards

Note:

  • Actual cylinder pressure may vary due to loss from quality testing procedure

Specifications

Solution Contains 65 components
Concentration 100 ppb
Solvent or Matrix Nitrogen
Quantity 110 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.