Learn More
Bioassay Systems EnzyChrom™ Monoamine Oxidase Assay Kit. For rapid, quantitative determination of monoamine oxidase activity and MAO inhibitor screen.
SDP
Catalog No. 501956317 Shop All Bioassay Systems Products
Encompass_Preferred
For rapid, quantitative determination of monoamine oxidase activity and MAO inhibitor screen. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.01 U/L MAO activity can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585nm. Samples: MAO enzyme preparations. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.01 U/L.
Catalog No. 50-195-6317 Supplier Bioassay Systems Supplier No. EMAO100
For rapid, quantitative determination of monoamine oxidase activity and MAO inhibitor screen. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.01 U/L MAO activity can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585nm. Samples: MAO enzyme preparations. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.01 U/L.