Learn More
Bioassay Systems EnzyChrom™ Monoamine Oxidase Inhibitor Screening Kit. HTS for inhibitor screening and evaluation of monoamine oxidase inhibitors.
SDP
Catalog No. 501956312 Shop All Bioassay Systems Products
Encompass_Preferred
For evaluation and high-throughput screen (HTS) of monoamine oxidase modulators. Key Features: Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585nm. Samples: Monoamine Oxidase. Species: All. Procedure: Assay takes 35 min. Kit size: 100 tests.
Catalog No. 50-195-6312 Supplier Bioassay Systems Supplier No. EIMO100
For evaluation and high-throughput screen (HTS) of monoamine oxidase modulators. Key Features: Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585nm. Samples: Monoamine Oxidase. Species: All. Procedure: Assay takes 35 min. Kit size: 100 tests.