Learn More
Bioassay Systems QuantiChrom™ Glutathione S-transferase Assay Kit. Glutathione S-transferase activity determination in biological samples.
SDP
Catalog No. 501956289 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of glutathione S-transferase activity and evaluation of drug modulators. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 2 to 80 U/L for 10 min reaction at 25°C. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD340nm. Samples: Biological samples (e.g. cell lysates, tissues, etc.). Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 2 U/L.
Catalog No. 50-195-6289 Supplier Bioassay Systems Supplier No. DGST100
For quantitative determination of glutathione S-transferase activity and evaluation of drug modulators. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 2 to 80 U/L for 10 min reaction at 25°C. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD340nm. Samples: Biological samples (e.g. cell lysates, tissues, etc.). Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 2 U/L.