Learn More
Bioassay Systems QuantiChrom™ Carbonyl Assay Kit. Quantification of carbonyl groups or protein carbonyls in biological samples.
SDP
Catalog No. 501956283 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of carbonyl groups or protein carbonyls and evaluation of drug modulators. Key Features: Fast and sensitive. Use of 100 µL sample. Linear detection range from 12 to 250 µM carbonyl in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 30 min. High-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD375nm. Samples: Chemicals with carbonyl groups (e.g. ketones, aldehydes) or protein carbonyls in biological samples (e.g. oxidized BSA, etc). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 12 µM.
Catalog No. 50-195-6283 Supplier Bioassay Systems Supplier No. DCAR100
For quantitative determination of carbonyl groups or protein carbonyls and evaluation of drug modulators. Key Features: Fast and sensitive. Use of 100 µL sample. Linear detection range from 12 to 250 µM carbonyl in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 30 min. High-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD375nm. Samples: Chemicals with carbonyl groups (e.g. ketones, aldehydes) or protein carbonyls in biological samples (e.g. oxidized BSA, etc). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 12 µM.