Learn More
Bioassay Systems QuantiChrom™ Urea Assay Kit II. For quantitative enzymatic determination of urea.
SDP
Catalog No. 501956294 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative enzymatic determination of urea. Key Features: Fast and sensitive. Linear detection range (20 ™L sample): 1 mg/mL (0.17 mM) to 100 mg/mL (17 mM) urea in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 5 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. Homogeneous "mix-incubate-measure" type assay.Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD557nm; Samples: Urea in biological samples (e.g. plasma, serum, urine) and food/beverage samples (e.g. milk). Size: 100 tests.
Catalog No. 50-195-6294 Supplier Bioassay Systems Supplier No. DUR2100
For quantitative enzymatic determination of urea. Key Features: Fast and sensitive. Linear detection range (20 ™L sample): 1 mg/mL (0.17 mM) to 100 mg/mL (17 mM) urea in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 5 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. Homogeneous "mix-incubate-measure" type assay.Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD557nm; Samples: Urea in biological samples (e.g. plasma, serum, urine) and food/beverage samples (e.g. milk). Size: 100 tests.