Learn More
Bioassay Systems EnzyChrom™ Malate Dehydrogenase Assay Kit. For quantitative determination of L-malate dehydrogenase enzyme activity and drug effects on malate dehydrogenase metabolism.
SDP
Catalog No. 501956318 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of L-malate dehydrogenase enzyme activity and drug effects on malate dehydrogenase metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.5 to 65 U/L for 20 min reaction at 37°C. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Biological samples (e.g. plasma, serum, erythrocytes, tissue and culture media). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.5 U/L.
Catalog No. 50-195-6318 Supplier Bioassay Systems Supplier No. EMDH100
For quantitative determination of L-malate dehydrogenase enzyme activity and drug effects on malate dehydrogenase metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.5 to 65 U/L for 20 min reaction at 37°C. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Biological samples (e.g. plasma, serum, erythrocytes, tissue and culture media). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.5 U/L.