Learn More
Bioassay Systems QuantiChrom™ G6PDH Inhibitor Screening Kit. HTS for inhibitor screening and evaluation of glucose-6-phosphate dehydrogenase inhibitors.
SDP
Catalog No. 501956331 Shop All Bioassay Systems Products
Encompass_Preferred
For evaluation and high-throughput screen (HTS) of glucose-6-phosphate dehydrogenase modulators. Key Features: Safe. Non-radioactive assay. High-throughput. Homogenous “mix-incubate-measure” type assay. Can be readily automated on HTS liquid handling system. Rapid and reliable. Can be completed in 30 minutes and no 37°C heater is needed. Method: OD565nm. Samples: Glucose-6-phosphate dehydrogenase. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests.
Catalog No. 50-195-6331 Supplier Bioassay Systems Supplier No. IGPDH100
For evaluation and high-throughput screen (HTS) of glucose-6-phosphate dehydrogenase modulators. Key Features: Safe. Non-radioactive assay. High-throughput. Homogenous “mix-incubate-measure” type assay. Can be readily automated on HTS liquid handling system. Rapid and reliable. Can be completed in 30 minutes and no 37°C heater is needed. Method: OD565nm. Samples: Glucose-6-phosphate dehydrogenase. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests.