Learn More
Bioassay Systems EnzyChrom™ Glycolysis Assay Kit. Direct determination of L-Lactate produced by glycolysis in cell samples.
SDP
Catalog No. 501956297 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of L-Lactate and screening for glycolysis modulators. Key Features: Fast and sensitive. Use of 5 µL sample. Linear detection range up to 10 mM L-lactate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Cell media. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests.
Catalog No. 50-195-6297 Supplier Bioassay Systems Supplier No. ECGL100
For quantitative determination of L-Lactate and screening for glycolysis modulators. Key Features: Fast and sensitive. Use of 5 µL sample. Linear detection range up to 10 mM L-lactate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Cell media. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests.