Learn More
Bioassay Systems EnzyChrom™ Isocitrate Assay Kit. For quantitative determination of isocitrate and evaluation of drug effects on its metabolism.
SDP
Catalog No. 501956298 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative colorimetric determination of isocitrate and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 20 to 5000 µM for 10 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Food, beverage, biological samples (e.g. cell lysate, tissue homogenate, serum, etc.). Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 20 µM.
Catalog No. 50-195-6298 Supplier Bioassay Systems Supplier No. ECIC100
For quantitative colorimetric determination of isocitrate and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 20 to 5000 µM for 10 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Food, beverage, biological samples (e.g. cell lysate, tissue homogenate, serum, etc.). Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 20 µM.