Learn More
Bioassay Systems EnzyChrom™ Formate Assay Kit. Direct determination of formate concentrations in biological (e.g. urine, blood, etc.) and other samples.
SDP
Catalog No. 501956305 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of formate and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Use of 10 µL sample. Linear detection range 0.050 to 5 mM Formate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 60 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Urine, Serum etc. Species: All. Procedure: Assay takes approximately 60 min. Kit size: 100 tests. Detection limit: 50 µM.
Catalog No. 50-195-6305 Supplier Bioassay Systems Supplier No. EFOR100
For quantitative determination of formate and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Use of 10 µL sample. Linear detection range 0.050 to 5 mM Formate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 60 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Urine, Serum etc. Species: All. Procedure: Assay takes approximately 60 min. Kit size: 100 tests. Detection limit: 50 µM.