Learn More
Bioassay Systems QuantiChrom™ Aldehyde Dehydrogenase Inhibitor Screening Kit. HTS for inhibitor screening and evaluation of aldehyde dehydrogenase inhibitors.
SDP
Catalog No. 501956311 Shop All Bioassay Systems Products
Encompass_Preferred
For evaluation and high-throughput screen (HTS) of aldehyde dehydrogenase modulators. Key Features: Rapid and reliable. Can be completed in under an hour. Homogenous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Aldehyde dehydrogenase. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests.
Catalog No. 50-195-6311 Supplier Bioassay Systems Supplier No. EIAL100
For evaluation and high-throughput screen (HTS) of aldehyde dehydrogenase modulators. Key Features: Rapid and reliable. Can be completed in under an hour. Homogenous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Aldehyde dehydrogenase. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests.