Learn More
Bioassay Systems EnzyChrom™ Invertase Assay Kit. For quantitative determination of invertase/sucrase activity.
SDP
Catalog No. 501956314 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of invertase/sucrase enzyme activity. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.007 U/L invertase activity can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological, environment (soil) etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.007 U/L.
Catalog No. 50-195-6314 Supplier Bioassay Systems Supplier No. EIVT100
For quantitative determination of invertase/sucrase enzyme activity. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.007 U/L invertase activity can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological, environment (soil) etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.007 U/L.