Learn More
Bioassay Systems EnzyChrom™ Aspartate Assay Kit. For quantitative determination of aspartate and evaluation of drug effects on aspartate metabolism.
SDP
Supplier:  Bioassay Systems EASP100
Encompass_Preferred
For quantitative determination of aspartate and evaluation of drug effects on aspartate metabolism. Key Features: Sensitive and accurate. Linear detection range: 2 to 400 µM aspartate for colorimetric assays and 1 to 50 µM for fluorimetric assays. Method: OD570nm, or FL530/585nm. Samples: Plasma, serum, tissue, culture media etc. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: OD, FL: 2, 1 µM.
Catalog No. 50-195-6296