Learn More
Polylc Inc 100 X 4.6 MM ID COLUMN 3UM 3
SDP
Supplier:  Polylc Inc 104SE0303
Encompass
Column; PolySulfoethyl A; 3um; 4.6 x 100mm; 300angstrom; Strong Cation-Exchange of Peptides
Catalog No. NC9183244