Learn More
Sino Biological HER2/ERBB2 Antibody (PE), Mouse MAb 25Tests
Supplier:  Sino Biological 10004MM07TP25
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells. Then conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-0342
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.