Learn More
Sino Biological Human CD112 / Nectin-2 / PVRL2 Protein 100ug
Supplier:  Sino Biological 10005HCCH100
Encompass_Preferred
The mature form of recombinant human CD112 (NP_002847.1) extracellular domain (Met 1-Leu 360) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-3758
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.