Learn More
Teknova 1000x Trace Metals Mixture. 1000mL, Sterile. 8/UN1760/III - 1L Limited
SDP
Supplier:  Teknova T1001
Encompass_Preferred
13.52mg/mL Ferric Chloride,2.94mg/mL Calcium Chloride,1.98mg/mL Manganese Chloride,2.88mg/mL Zinc Sulfate,476µg/mL Cobalt Chloride,341µg/mL Cupric Chloride,455µg/mL Nickel Chloride,489µg/mL Sodium Molybdate,346µg/mL Sodium Selenite,124µg/mL Boric Acid
Catalog No. NC0112668
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.