Learn More
Sino Biological GDF3 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 100344T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human GDF3, and purified by antigen affinity chromatography.
Catalog No. 50-161-0925
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.