Learn More
Sino Biological Human ANPEP / CD13 Protein (603 Met/Ile, His Tag) 50ug
Supplier:  Sino Biological 10051H08H50
Encompass_Preferred
A DNA sequence encoding the metalloprotease domain (Lys 69-Lys 967,603 Met/Ile) of the mature form of human ANPEP (NP_001141.2) was expressed with N-terminal signal peptide and C-terminal polyhistidine tag.
Catalog No. 50-161-3885
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.