Learn More
Sino Biological Human PD-L1 / B7-H1 / CD274 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10084H02H100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Thr 239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-3960
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.