Learn More
Sino Biological Human IL-8 / CXCL8 Protein (aa 23-99, Fc Tag) 100ug
Supplier:  Sino Biological 10098H01H1100
Encompass_Preferred
A DNA sequence encoding the 77 amino acid endothelial-cell derived form ofthe mature human IL8 (NP_000575.1) (Ala 23-Ser 99) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-3981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.