Learn More
Clontech Labs 3P 100BP LADDER 500UL
SDP
Supplier:  Clontech Labs 3P 3407A
Encompass
DNA Marker; Clontech; TaKaRa; Used as molecular weight standard; 100bp DNA ladder; Fragments: 100bp to 1kb, additional 1.5kb; Ten bands; 100ng/uL; For 100 loadings; Premied w/6X loading buffer for direct gel electrophoresis; Size: 500ug/mL
Catalog No. 50-443-772
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.