Learn More
The Fresh Test 100g Glucose Powder 60/pk
Supplier:  The Fresh Test 2710205760BOX
Encompass
The Fresh Test 100g Glucose Powder is a diagnostic glucose beverage intended for use in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each pouch contains 100 grams of glucose to be reconstituted with 10 oz. of water prior to administration. For an optimal experience, the beverage should be consumed cold. When used for a 100-gram glucose load test, the beverage should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
• Non-GMO Dextrose, Crystallized Lemon, Organic Mint
• 30-month shelf life
• BPA-free pouch
• Kosher Certified; Non-GMO Project Verified
• Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 60 units per box.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.
Catalog No. NC3627355