Learn More
SCP Science CONOSTAN 100ppm Standard, Hg (75cSt)
Supplier:  SCP Science 150101801
Encompass
Single element, metallo-organic standards blended in 75 cSt blank oil. Complete with a Certificate of Analysis. For use with ICP, AA, rotrode, XRF, DCP, flame emission and other instruments.
Catalog No. NC1327625
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.