Learn More
ILEX LIFE SCIENCES LLC 100X NEUROPAN-2 100 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0711100
Encompass_Preferred
PAN-Biotech Neuropan-2 Supplement (100x): Serum-Free Supplement for Cultivation of Neurons (100 ml). DESCRIPTION: Neuropan-2 is a supplement formulated to meet special requirements of neuronal cells. When supplemented with Neuropan Basal Medium, it allows long-term maintenance of normal phenotype and growth of neuronal cells, and maintains pure populations of neuronal cells without need for an astrocyte feeder. Neuropan-2 meets the special cell culture requirements of prenatal and embryonic neuronal cells from hippocampus, cortex, and other regions of the brain. APPLICATION: Neuropan-2 Supplement is a chemically-defined, serum-free supplement for the growth of primary embryonic neurons as well as embryonic neurospheres/CNS progenitor cells when combined with bFGF and EGF in Neuropan Basal Medium. COMPOSTION: Neuropan-2 Supplement contains insulin, transferrin, albumin, and hormones. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3708
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.