Learn More
Sino Biological CD55/DAF Antibody (APC), Mouse MAb 100Tests
Supplier:  Sino Biological 10101MM04A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD55/DAF (rh CD55/DAF; Catalog#10101-H08H; NP_000565.1; Met1-Ser353) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-2666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.