Learn More
Sino Biological Human IL36G / IL1F9 Protein 50ug
Supplier:  Sino Biological 10124HNAE50
Encompass_Preferred
A DNA sequence encoding the human IL1F9 isoform 1 (Q9NZH8-1) (Met 1-Asp 169) was expressed.
Catalog No. 50-161-4074
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.