Learn More
Sino Biological CTSZ / CTSX / Cathepsin Z Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10159MM05100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CTSZ / CTSX / Cathepsin Z (rh CTSZ / CTSX / Cathepsin Z; Catalog#10159-H08H; Q9UBR2; Met1-Val303). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2163