Learn More
Sino Biological Recombinant Human NBL1 Protein (hFc Tag)
Supplier:  Sino Biological 10169H02H100
Encompass_Preferred
A DNA sequence encoding the human DAN precursor (NP_005371.1) (Met 1-Asp 180) was expressed with C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4197
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.