Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclohexanemethylamine

Catalog No. 1018425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-184-25G 5 g
10-184-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-184-25G Supplier Sigma Aldrich Supplier No. 1018425G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclohexanemethylamine was used in the synthesis of schiffαs base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(ii) complexes
  • Cyclohexanemethylamine was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs

Specifications

Density 0.87 g/mL (at 25°C (literature))
Boiling Point 159°C to 161°C (lit.)
Molecular Formula C7H15N
Refractive Index n20/D 1.462 (literature)
Linear Formula C6H11CH2NH2
MDL Number MFCD00001520
Quantity 5 g
Synonym (Aminomethyl)cyclohexane; Hexahydrobenzylamine
Molecular Weight (g/mol) 113.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.