Learn More
Sino Biological Human ESAM / Endothelial Cell Adhesion Molecule Protein 100ug
Supplier:  Sino Biological 10187HCCH100
Encompass_Preferred
The mature form of human ESAM (NP_620411.2) extracellular domain (Met 1-Ala 248) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.