Learn More
Sino Biological Recombinant Human HER3/ERBB3 Protein (mFc Tag)
Supplier:  Sino Biological 10201H05H100
Encompass_Preferred
A DNA sequence encoding the human ErbB3 (NP_001973.2) (Met1-Thr643)was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-4302
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.