Learn More
Sino Biological Human cytomegalovirus (HCMV) Glycoprotein B / gB Protein 100ug
Supplier:  Sino Biological 10202VCCH1100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907) of human CMV gB (AAA45920.1, with furin cleavage site mutated from 'RTKR' to 'TTQT') was expressed, fused with five additional amino acids (DDDDK) at the C-terminus.
Catalog No. 50-161-4318
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.